missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RoboSep™ 'Do-It-Yourself' Positive Selection Kit II
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
1 set
Description
- The EasySep™ 'Do-It-Yourself' Selection Kit II is designed to select any cell type of interest with your own mouse IgG1 monoclonal antibody
- Cells that express the surface antigen that is recognized by the mouse IgG1 monoclonal antibody are targeted for magnetic separation using EasySep™ Dextran RapidSpheres™ and the EasySep™ Magnet
- This product replaces the EasySep™ 'Do-It-Yourself' Selection Kit (Catalog #18098)
Specifications
Specifications
| Cell Type | B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction