missing translation for 'onlineSavingsMsg'
Learn More

Robenidine Hydrochloride, TRC

Product Code. 17945025
Change view
Click to view available options
Quantity:
100 mg
10 g
100 g
Unit Size:
100g
100mg
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17945025 100 mg 100mg
30134607 10 g 10g
30134330 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17945025 Supplier Toronto Research Chemicals Supplier No. TRCR639000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25875-50-7
Formula Weight 369.0315
InChI Formula InChI=1S/C15H13Cl2N5.ClH/c16-13-5-1-11(2-6-13)9-19-21-15(18)22-20-10-12-3-7-14(17)8-4-12;/h1-10H,(H3,18,21,22);1H/b19-9+,20-10+;
IUPAC Name 1,3-bis[(E)-(4-chlorophenyl)methylideneamino]guanidine;hydrochloride
Molecular Formula C15 H13 Cl2 N5 . Cl H
Molecular Weight (g/mol) 370.66
SMILES Cl.Clc1ccc(\C=N\NC(=N)N\N=C\c2ccc(Cl)cc2)cc1
Synonym 2,2'-Bis[(4-chlorophenyl)methylene]carbonimidic Dihydrazide Hydrochloride (1:1), Carbonimidic dihydrazide, bis[(4-chlorophenyl)methylene]-, monohydrochloride (9CI), Guanidine, 1,3-bis[(p-chlorobenzylidene)amino]-, monohydrochloride (8CI), 1,3-Bis[(p-chlorobenzylidene)amino]guanidine monohydrochloride, Cycostat, NCL 812, Robenidine hydrochloride, Robenzidene
Chemical Name or Material Robenidine hydrochloride

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.