Learn More
Ro 48-8071 fumarate, MedChemExpress
Product Code. 16493567
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16493567
Brand: Medchem Express HY18630A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Ro 48-8071 fumarate is an inhibitor of OSC (Oxidosqualene cyclase) with IC50 of appr 6.5 nM.
Ro 48-8071 fumarate is an inhibitor of OSC (Oxidosqualene cyclase) with IC50 of appr 6.5 nM.
Specifications
Chemical Name or Material Ro 48-8071 fumarate
CAS 189197-69-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H31BrFNO6
Quantity 50 mg
Solubility Information H2O : 100 mg/mL (177.17 mM; Need ultrasonic) ∣DMSO : ≥ 55 mg/mL (97.44 mM)
SMILES O=C(C1=CC=C(Br)C=C1)C2=CC=C(OCCCCCCN(C)CC=C)C=C2F.O=C(O)/C=C/C(O)=O
Molecular Weight (g/mol) 564.44
Formula Weight 564.44
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death