missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF24 Monoclonal Antibody (4C6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521310
This item is not returnable.
View return policy
Description
Peptide Sequence: LRHQAHKEFY AYKQVILKEK VKELNLHELC AVCLEDFKPR DELGICPCKH AFHRKCLIKW LEVRKVCPLC NMPVLQLAQL HSKQDRGPPQ GPLPGAENIV.
The protein encoded by this gene contains similarity to the Drosophila goliath protein and thus may function as a transcription factor. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene. [provided by RefSeq, Aug 2008].
Specifications
| RNF24 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9Y225 | |
| RNF24 | |
| RNF24 (NP_009150, 49 a.a. ~ 148 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Immunocytochemistry | |
| 4C6 | |
| Unconjugated | |
| RNF24 | |
| 2810473M14Rik; 4930505A13Rik; AI317164; C86507; D2Ertd504e; G1L; RING finger protein 24; Rnf24 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11237 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction