missing translation for 'onlineSavingsMsg'
Learn More

RNF150 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16821035 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16821035 150 μL 150µL
16831035 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16821035 Supplier Proteintech Supplier No. 214381AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNF150 contains a RING-type zinc finger, a motif known to be involved in protein-protein and protein-DNA interactions. The specific function of this protein is unknown.

Specifications

Antigen RNF150
Applications Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RNF150
Gene Accession No. Q5DTZ6, Q9ULK6
Gene Alias KIAA1214, ring finger protein 150, RNF150
Gene Symbols Rnf150
Host Species Rabbit
Immunogen RNF150 Fusion Protein Ag16064
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 330812, 364983, 57484
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.