Learn More
Beckman Coulter RNAdvance Cell V2 Total RNA Isolation Kit
Product Code. 31723149
Change view
Click to view available options
No. of Reactions:
100 Preparations
960 Preparations
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
31723149 960 Preparations 1 set
31723128 100 Preparations 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Refer to SDS
Product Code. 31723149 Supplier Beckman Coulter Supplier No. A47943
Please to purchase this item. Need a web account? Register with us today!
Refer to SDS
Based on patented Solid Phase Reversible Immobilization (SPRI) paramagnetic bead technology, the RNAdvance Cell v2 Kit is known for consistently delivering pure nucleic acids of high quality and yield.
  • This system reliably delivers high RNA recovery and purity without the need for filtration or centrifugation.
  • The Kit chemistry is an automation-friendly process for isolating total RNA for utilization in microarray and real time qPCR* applications.
*The PCR process is covered by patents owned by Roche Molecular Systems, Inc., and F. Hoffman-La Roche, Ltd.
TRUSTED_SUSTAINABILITY
Specifications
Sample Type Cells
No. of Reactions 960 Preparations
Product Type RNA Extraction Kit