Learn More
Beckman Coulter RNAdvance Cell V2 Total RNA Isolation Kit
Description
- This system reliably delivers high RNA recovery and purity without the need for filtration or centrifugation.
- The Kit chemistry is an automation-friendly process for isolating total RNA for utilization in microarray and real time qPCR* applications.
*The PCR process is covered by patents owned by Roche Molecular Systems, Inc., and F. Hoffman-La Roche, Ltd.
Specifications
Specifications
| Sample Type | Cells |
| No. of Reactions | 100 Preparations |
| Product Type | RNA Extraction Kit |