Learn More
Risedronic Acid-d4 (Major), TRC
Product Code. 30409448
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409448 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30409448 Supplier Toronto Research Chemicals Supplier No. TRCR52150210MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Risedronic Acid-d4
CAS 1035438-80-2
Molecular Formula C7H7D4NO7P2
Quantity 10 mg
Synonym [1-Hydroxy-2-(3-pyridinyl)ethylidene]bis phosphonic Acid-d4, Actonel-d4, NE 58095-d4
InChI Formula InChI=1S/C7H11NO7P2/c9-7(16(10,11)12,17(13,14)15)4-6-2-1-3-8-5-6/h1-3,5,9H,4H2,(H2,10,11,12)(H2,13,14,15)/i1D,2D,3D,5D
SMILES O=P(O)(O)C(P(O)(O)=O)(O)CC1=C([2H])C([2H])=C([2H])N=C1[2H]
Molecular Weight (g/mol) 287.14
Formula Weight 287.0262