missing translation for 'onlineSavingsMsg'
Learn More

RIP140 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

189.00 €

Specifications

Antigen RIP140
Concentration 1 mg/mL
Dilution Western Blot 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18577350
View Documents
Novus Biologicals
NB110-5122-0.025mg
0.025 mg
189.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RIP140 Polyclonal specifically detects RIP140 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

RIP140
Western Blot 2 μg/mL, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Polyclonal
Rabbit
Transcription Factors and Regulators
PBS, 1 mg/ml BSA with 0.05% Sodium Azide
8204
Synthetic peptide corresponding to residues G(718) N P T K G R V K K K E K T P L R D(735) of human RIP140.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
140 kDa
1 mg/mL
Western Blot
Unconjugated
RUO
Human
10RIP140Receptor-interacting protein 140, FLJ77253, nuclear receptor interacting protein 1, nuclear receptor-interacting protein 1, receptor interacting protein 140
NRIP1
IgG
Affinity Purified
Detects a band at approximately 140 kDa.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.