missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIOK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97641-50ul
This item is not returnable.
View return policy
Description
RIOK2 Polyclonal antibody specifically detects RIOK2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| RIOK2 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| EC 2.7.11.1, FLJ11159, RIO kinase 2, RIO kinase 2 (yeast), RIO2, serine/threonine-protein kinase RIO2 | |
| Produced in rabbits immunized with E. coli-derived Human RIOK2 fragment. | |
| 50 μg | |
| Protein Kinase | |
| 55781 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction