missing translation for 'onlineSavingsMsg'
Learn More

Riociguat, MedChemExpress

Product Code. 16471136
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16471136

Brand: Medchem Express HY14779

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Riociguat is an oral stimulator of soluble guanylate cyclase (sGC) used in the treatment of pulmonary hypertension.

Riociguat is an oral stimulator of soluble guanylate cyclase (sGC) used in the treatment of pulmonary hypertension.

Specifications

Chemical Name or Material Riociguat
CAS 625115-55-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H19FN8O2
Quantity 200 mg
Synonym BAY 632521
Solubility Information DMSO : 20 mg/mL (47.35 mM; Need ultrasonic)
SMILES FC1=CC=CC=C1CN2C3=NC=CC=C3C(C4=NC(N)=C(C(N)=N4)N(C)C(OC)=O)=N2
Molecular Weight (g/mol) 422.42
Formula Weight 422.42
Percent Purity 95.1%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.