Learn More
Rimantadine-d4 Hydrochloride, TRC
Product Code. 30421506
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421506 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30421506 Supplier Toronto Research Chemicals Supplier No. TRCR5170022.5MG
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Rimantadine-d4 Hydrochloride
CAS 350818-67-6
Purity Grade Notes HPLC
Molecular Formula C12 D4 H17 N . Cl H
Quantity 2.5 mg
Synonym Tricyclo[3.3.1.13,7]decane-1-methan-α-d-amine','α-(methyl-d3)-','hydrochloride (9CI), α-(Methyl-d3)tricyclo[3.3.1.13,7]decane-1-methan-α-d-amine hydrochloride, (±)-Rimantadine-d4 hydrochloride, 1-(1-Adamantyl)ethylamine-1,2,2,2-d4 hydrochloride, 1-(1-Aminoethyl-1,2,2,2-d4)adamantane hydrochloride, 1-(Adamantan-1-yl)ethanamine-d4 hydrochloride, 1-Rimantadine-d4 hydrochloride, Rimantadine-d4 hydrochloride, Rimantadine-D4 (aminoethyl-1,2,2,2-D4) hydrochloride
InChI Formula InChI=1S/C12H21N.ClH/c1-8(13)12-5-9-2-10(6-12)4-11(3-9)7-12;/h8-11H,2-7,13H2,1H3;1H/i1D3,8D;
SMILES Cl.[2H]C([2H])([2H])C([2H])(N)C12CC3CC(CC(C3)C1)C2
IUPAC Name 1-(1-adamantyl)-1,2,2,2-tetradeuterioethanamine;hydrochloride
Molecular Weight (g/mol) 219.787
Formula Weight 219.169
Percent Purity >95