missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RIM1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5140003

Product Code. 17844520

  • 464.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ENDRHSRKSE RSSIQKQTRK GTASDAERVL PTCLSRRGHA APRATDQPVI RGKHPARSRS SEHSSIRTLC SMHHL Highest antigen sequence indentity to the following orthologs: Rat - 85%, Mouse - 91%.

Rims1 is a RAS gene superfamily member that regulates synaptic vesicle exocytosis. Rims1 also plays a role in the regulation of voltage-gated calcium channels during neurotransmitter and insulin release. Mutations in the Rims1 gene have suggested a role cognition and have been identified as the cause of cone-rod dystrophy type 7. Further, Rims1 may act as scaffold protein that regulates neurotransmitter release at the active zone. Rims1 may also be essential for maintaining normal probability of neurotransmitter release and for regulating release during short-term synaptic plasticity. Rims1 also plays a role in dendrite formation by melanocytes. Multiple transcript variants encoding different isoforms have been described for this Rims1. Diseases associated with RIMS1 include Cone-Rod Dystrophy 7 and Retinitis Pigmentosa.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RIM1
Polyclonal
Unconjugated
Rims1
C030033M19Rik; CORD7; Kiaa0340; mKIAA0340; Nbla00761; Rab3 effector; Rab3 interacting protein 1; Rab-3-interacting molecule 1; rab3-interacting molecule 1; rab-3-interacting protein 1; rab3-interacting protein 1; Rab-3-interacting protein 2; RAB3-interacting protein 2; Rab3ip1; RAB3IP2; regulating synaptic membrane exocytosis 1; regulating synaptic membrane exocytosis 1 beta; regulating synaptic membrane exocytosis protein 1; Rim; RIM 1; RIM1; RIM1a; RIM1alpha; Rim1b protein; Rims1; Serg1; synaptic exocytosis regulator 1
Rabbit
Antigen affinity chromatography
RUO
22999
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q86UR5
Rims1
Recombinant protein corresponding to Human RIMS1. Recombinant protein control fragment (Product #RP-109568).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.