Learn More
Atrow™ Rilsan Hanging Rack
Product Code. 10406143
Change view
Click to view available options
Holds:
Tubes and Flasks
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
10406143 Tubes and Flasks Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10406143 Supplier Atrow™ Supplier No. 899114
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Holds laboratory glassware for efficient air drying. Atrow™ Rilsan Hanging Rack is a wall-mounted rack that can dry dozens of tubes and flasks simultaneously without occupying counter space. It is made of 6mm stainless steel rods coated with Rislan™ for excellent mechanical and chemical resistance.
Rilsan™ is a biobased polymer resin coating with outstanding impact, durability and chemical resistance properties.
  • 9 double pins for holding flasks
  • 53 single pin for suspending tubes
  • Size: 45cmL x 54cmH
Specifications
Color White
Holds Tubes and Flasks
Material Stainless Steel, Rilsan
No. per Pack 1 Pack
Quantity 1 Rack
Length (Metric) 450 mm
Product Type Wall Mounted Drainer
Height (Metric) 540 mm