Learn More
Rilmenidine Hemifumarate, TRC
Product Code. 17990455
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990455 10 mg 10mg
17932305 1 mg 1mg
17911635 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17990455 Supplier Toronto Research Chemicals Supplier No. TRCR50970510MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 207572-68-7
Molecular Formula 2 C10 H16 N2 O . C4 H4 O4
Molecular Weight (g/mol) 476.566
Synonym N-(Dicyclopropylmethyl)-4,5-dihydro-2-oxazolamine (E)-2-Butenedioate, S 3341, Oxaminozoline;
IUPAC Name (E)-but-2-enedioic acid;N-(dicyclopropylmethyl)-4,5-dihydro-1,3-oxazol-2-amine
SMILES OC(=O)\C=C\C(=O)O.C1CN=C(NC(C2CC2)C3CC3)O1.C4CN=C(NC(C5CC5)C6CC6)O4
Specifications
Quantity 10 mg
InChI Formula InChI=1S/2C10H16N2O.C4H4O4/c2*1-2-7(1)9(8-3-4-8)12-10-11-5-6-13-10;5-3(6)1-2-4(7)8/h2*7-9H,1-6H2,(H,11,12);1-2H,(H,5,6)(H,7,8)/b;;2-1+
Formula Weight 476.263
Chemical Name or Material Rilmenidine Hemifumarate