Learn More
Brady™ Rigid Polypropylene Prohibition Signs
Product Code. 11840762
Click to view available options
Displayed Text:
Prohibits persons with a pacemaker
Standing and Parking Prohibited
Unit Size:
Each
Product Code. 11840762
Brand: Brady™ 252052
Please to purchase this item. Need a web account? Register with us today!
Specifications
Alternate Part Number PIC 226-DIA 200-B7527
Material Polypropylene
Quantity 1 Ea.
Description Rigid Polypropylene
Diameter (Metric) 200 mm
For Use With (Application) Used for a wide variety of applications both indoors and outdoors
Thickness 0.158 mm
Width (Metric) 105 mm
Shape Round
Displayed Text Standing and Parking Prohibited
Product Type Prohibition Sign
Color Red