Learn More
Brady™ Rigid Polypropylene Extinguisher Sign: No Water Pictogram
Product Code. 10575694 Shop All Brady Products
Change view
Click to view available options
Diameter (Metric):
200 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10575694 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10575694 Supplier Brady™ Supplier No. 252009
  • ONLINE EXCLUSIVE PRICE
    13.60 €
    12.26 € / Pack of 1
    Save 1.34 €
    9% Off
Estimated Shipment: 17-11-2026