missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RIG-I Recombinant Rabbit Monoclonal Antibody (2J8D5)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA555547

Product Code. 30131191

  • 457.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen Sequence: FKPRNYQLEL ALPAMKGKNT IICAPTGCGK TFVSLLICEH HLKKFPQGQK GKVVFFANQI PVYEQQKSVF SKYFERHGYR VTGISGATAE NVPVEQIVEN NDIIILTPQI LVNNLKKGTI PSLSIFTLMI FDECHNTSKQ HPYNMIMFNY LDQKLGGSSG PLPQVIGLTA SVGVGDAKNT DEALDYICKL CASLDASVIA TVKHNLEELE QVVYK.

Retinoic acid-inducible gene I, RIG-I is a pattern recognition receptor (PRR) involved in the recognition of viral dsRNA. Along with MDA5, RIG-I detects viral dsRNA and activates the innate immune response. Both MDA5 and RIG-I are RNA helicases and they perform overlapping as well as distinct roles. RIG-I is activated by dsRNAs without a 5'-triphosphate end and short dsRNAs, whereas MDA5 is activated by long dsRNAs. Once activated, both proteins signal through IPS-1 activating transcription factors NF-kappaB and IRF-3 (1) and ultimately activating apoptosis, cytokine signaling, and inflammation. RIG-I is essential for signaling by influenza A, influenza B, human respiratory syncytial virus (3), paromyxoviruses, Japanese encephalitis virus, and West Nile virus. MicroRNA-146a has been implicated in feedback inhibition of RIG-I-dependant antiviral response by negatively regulating RIG-I targets TRAF6, IRAK1, and IRAK2. Recent evidence has implicated RIG-I in the detection of cytosolic DNA through RNA polymerase III activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RIG-I
Recombinant Monoclonal
1.1 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
O95786, Q6Q899
DDX58
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse
Antibody
IgG
ELISA, Western Blot
2J8D5
Unconjugated
DDX58
6430573D20Rik; C330021E21; Ddx58; DEAD (Asp-Glu-Ala-Asp) box polypeptide 58; DEAD box protein 58; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide; DEAD/H box polypeptide RIG-I; DEAD-box protein 58; DEXD/H-box helicase 58; probable ATP-dependent RNA helicase DDX58; Retinoic acid-inducible gene 1 protein; retinoic acid-inducible gene I protein; retinoic acid-inducible gene-I; RIG-1; RIGI; RIG-I; RIG-I-like receptor 1; RLR-1; RNA helicase RIG-I; SGMRT2
Rabbit
Affinity chromatography
RUO
230073, 23586
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.