Learn More
Rifabutin-d7, TRC
Specifications
Specifications
| Chemical Name or Material | Rifabutin-d7 |
| CAS | 2747918-39-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C46H56D7N4O11 |
| Quantity | 0.5 mg |
| Synonym | 1',4-Didehydro-1-deoxy-1,4-dihydro-5'-(2-methylpropyl)-1-oxorifamycin XIV-d7, Ansatipine-d7, Ansamycin-d7, Antibiotic LM 427-d7, LM 427-d7, Mycobutin-d7, Rifabutine-d7 |
| InChI Formula | InChI=1S/C44H60N4O10/c1-21(2)20-48-17-15-44(16-18-48)46-32-29-30-38(52)27(8)40-31(29)41(54)43(9,58-40)57-19-14-28(56-10)24(5)36(50)26(7)37(51)25(6)35(49)22(3)12-11-13-23(4)42(55)45-34(39(30)53)33(32)47-44/h11-14,19,21-22,24-26,28,35-37,46,49-53H,15-18,20H2,1-10H3/b12-11+,19-14+,23-13+,45-34?/t22-,24+,25+,26-,28-,35-,36+,37+,43-/m0/s1/i1D3,2D3,21D |
| SMILES | O=C(NC1=C2C(C(C3=C4C(C)=C5O)=C5C1=O)=NC6(CCN(CC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])CC6)N2)/C(C)=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@H](C)[C@@H](OC)/C=C/O[C@](O4)(C)C3=O |
| Molecular Weight (g/mol) | 854.04 |
| Percent Purity | >95 |