missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RICTOR Monoclonal Antibody (J.644.6)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA515006
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
RICTOR and MTOR (FRAP1) are components of a protein complex that integrates nutrient- and growth factor-derived signals to regulate cell growth.
Specifications
| RICTOR | |
| Monoclonal | |
| 172 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q6QI06, Q6R327 | |
| Rictor | |
| Synthetic peptide corresponding to the sequence Gln1681 of human Rictor. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Western Blot | |
| J.644.6 | |
| Unconjugated | |
| Rictor | |
| 4921505C17Rik; 6030405M08Rik; AVO3; AVO3 homolog; AW492497; D530039E11Rik; DKFZp686B11164; Fyb; FYN binding protein; hAVO3; Kiaa1999; mAVO3; MGC39830; PIA; pianissimo; protein pianissimo; Rapamycin-insensitive companion of mTOR; rictor; RPTOR independent companion of MTOR complex 2; RPTOR independent companion of MTOR, complex 2; TORC2-specific protein AVO3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 253260, 310131, 78757 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction