Learn More
Epredia™ Richard-Allan Scientific™ Gram Stain (Tissue) Kit, Crystal Violet Solution
Shop All Epredia Products
Click to view available options
Type:
Safranin O Stain Solution
Unit Size:
500mL
Description
- Gram-positive organisms stain blue to blue-black
- Gram-negative organisms stain red
- Tartrazine provides a yellow background stain
Specifications
Specifications
| Type | Safranin O Stain Solution |
| Quantity | 500mL |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Specific target organ toxicity after single exposure Category 1
- H224-Extremely flammable liquid and vapour.
- H370-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : isopropyl alcohol, ethyl alcohol, methylalcohol, phenazinium, 3,7-diamino-2,8-dimethyl-5-phenyl chl