Learn More
Invitrogen™ Ribosome-inactivating protein Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598352
This item is not returnable.
View return policy
Description
Ribosome-inactivating protein Polyclonal Antibody for Western Blot, ELISA
Has cytotoxic activity towards cancer cells, but not normal cells. Inhibits the growth of the human leukemia cell line K562, the murine melanoma cell line B16 and the lung adenocarcinoma cell line A549 with IC50 values of 88.1 nM, 63.4 nM and 359.3 nM respectively. [Uniprot].
Specifications
| Ribosome-inactivating protein | |
| Polyclonal | |
| Unconjugated | |
| cucurmosin; Ribosome-inactivating protein cucurmosin; rRNA N-glycosidase | |
| Recombinant Cucurbita moschata Ribosome-inactivating protein cucurmosin protein (1-244AA). | |
| 100 μg | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Plant | |
| Antibody | |
| IgG |