missing translation for 'onlineSavingsMsg'
Learn More

Ribosomal Protein S6/RPS6 Antibody (522731), FITC, Novus Biologicals™

Product Code. 18622924 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18622924 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18622924 Supplier Novus Biologicals Supplier No. FAB5436F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ribosomal Protein S6/RPS6 Monoclonal antibody specifically detects Ribosomal Protein S6/RPS6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ribosomal Protein S6/RPS6
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 522731
Conjugate FITC
Dilution Western Blot, Immunocytochemistry
Formulation PBS
Gene Alias Phosphoprotein NP33, 40S ribosomal protein S6, pS6, ribosomal protein S6, S6
Host Species Mouse
Immunogen E. coli-derived recombinant human RPS6, Met1-Lys249, Accession # P62753
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, mTOR Pathway, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 6194
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.