missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riboflavin phosphate sodium, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Riboflavin phosphate sodium (FMN-Na) is a derivative of Riboflavin (vitamin B2) which is an essential nutrient for animals. Riboflavin phosphate sodium can be used for the research of progressive keratoconus, corneal ectasia and irregular astigmatism. Riboflavine phosphate sodium is a very effective NAD+-recycling agent.
Specifications
Specifications
| Chemical Name or Material | Riboflavin phosphate sodium |
| CAS | 130-40-5 |
| Color | Orange |
| Purity Grade Notes | Research |
| Molecular Formula | C17H20N4NaO9P |
| Quantity | 500 mg |
| Synonym | FMN-Na Riboflavin 5'-phosphate sodium Vitamin B2 Phosphate Sodium Salt |
| Solubility Information | H2O : 20.83 mg/mL (43.55 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 1 mg/mL (2.09 mM; Need ultrasonic) |
| SMILES | CC1=CC2=C(C=C1C)N=C(C(N2C[C@H](O)[C@H](O)[C@H](O)COP(O)(O[Na])=O)=N3)C(NC3=O)=O |
| Molecular Weight (g/mol) | 478.33 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction