missing translation for 'onlineSavingsMsg'
Learn More

Riboflavin-5'-phosphate sodium salt dihydrate

Product Code. p-7021984
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11491918

Brand: Thermo Scientific Alfa Aesar A14545.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 6184-17-4 | C17H24N4NaO11P | 514.36 g/mol

Riboflavin 5?-monophosphate (FMN) is used as a prosthetic group by oxidoreductase (one and two electron transfer) enzymes such as NADH dehydrogenase, nitric oxide synthase, and nitrilotriacetate monooxygenase. Riboflavin 5?-monophosphate is used as a substrate to study the specificity and kinetics of FMN phosphohydrolases and to study molecular sensing based gene regulation by riboswitches. It is a photoinitiator of polymerization for PAGE gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Riboflavin 5′-monophosphate (FMN) is used as a prosthetic group by oxidoreductase (one and two electron transfer) enzymes such as NADH dehydrogenase, nitric oxide synthase, and nitrilotriacetate monooxygenase. Riboflavin 5′-monophosphate is used as a substrate to study the specificity and kinetics of FMN phosphohydrolases and to study molecular sensing based gene regulation by riboswitches. It is a photoinitiator of polymerization for PAGE gels.

Solubility
Soluble in water 50 g/L @ 20°C.

Notes
Store in refrigerator. Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6184-17-4
Molecular Formula C17H24N4NaO11P
Molecular Weight (g/mol) 514.36
MDL Number MFCD00150993
InChI Key LQVGUKOCMOKKJU-FTNKLFHDNA-M
Synonym riboflavin 5'-monophosphate sodium salt dihydrate
PubChem CID 131856174
IUPAC Name [(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] dihydrogen phosphate;sodium;dihydrate
SMILES O.O.[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Melting Point >300°C
Quantity 10 g
Beilstein 4106529
Merck Index 14,8201
Solubility Information Soluble in water 50g/L @ 20°C.
Formula Weight 514.36 (478.34 Anhydrous)
Chemical Name or Material Riboflavin-5'-phosphate sodium salt dihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.