missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodopsin Antibody (1D4), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-47602-0.025mg
This item is not returnable.
View return policy
Description
Rhodopsin Monoclonal specifically detects Rhodopsin in Human, Mouse, Bovine, Vertebrate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Rhodopsin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| CSNBAD1, MGC138309, OPN2MGC138311, opsin 2, rod pigment, opsin-2, retinitis pigmentosa 4, autosomal dominant, rhodopsin, RP4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects ∼40kDa. Recognizes Rhodopsin (native and recombinant forms). No known reactivity to other proteins. Binds specifically to the C-terminal epitope -T-E-T-S-Q-V-A-P-A-(COOH). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 1D4 | |
| Unconjugated | |
| P08100 | |
| RHO | |
| Bovine Rhodopsin | |
| 0.025 mg | |
| GPCR, Signal Transduction, Vision | |
| 6010 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction