Learn More
Molecular Probes™ Rhodamine Green™ Carboxylic Acid, Succinimidyl Ester, Hydrochloride (5(6)-CR 110, SE), mixed isomers
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
This mixed isomer preparation of the amine-reactive Rhodamine Green carboxylic acid, succinimidyl ester yields bright and photostable conjugates. Rhodamine Green dye—labeled probes have been frequently used for Fluorescence Correlation Spectroscopy (FCS).
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Carboxylic Acid Succinimidyl Ester |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | Rhodamine Green™ |
| Label Type | Classic Dyes |
| Product Line | Rhodamine Green |