Learn More
Rheodyne™ Stainless-Steel Sample Loops
Product Code. 10686601
Click to view available options
Volume (Metric):
2 mL
This item is not returnable. View return policy
Product Code. 10686601
Brand: Rheodyne™ 702800
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
For manual sample injectors
Supplied with unswaged fittings. The two ends of the loop must be completely bottomed in the injector ports before the ferrule is swaged onto the loop. Swaging each end separately and then replacing the ends in their respective ports of the same valve ensure that the loop ends are bottomed into the ports. A fitting made up in one port may leave an undesirable cavity in another port. As all ports vary in all valves, careful attention to loop installation is important.
  • For 7125 and 7010 Sample Injectors (Cat. Nos. 06-658-1 and -2)
  • Factory-cut and finished to the highest quality
  • 316 stainless-steel loop ends have square cut and are burr-free, offering flush connection to valve
Specifications
Size (Metric) 2 mL
Volume (Metric) 2 mL