missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RGS10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106721
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RGS10.
RGS10 inhibits signal transduction by increasing the GTPASE activity of G protein alpha subunits thereby driving them into their inactive GDP-bound form. It associates specifically with the activated forms of the G protein subunits G(I)-ALPHA and G(Z)- alpha but fails to interact with the structurally and functionally distinct G(S)-alpha subunit. Activity on G(Z)-alpha is inhibited by palmitoylation of the G-protein.
Specifications
| RGS10 | |
| Polyclonal | |
| Unconjugated | |
| RGS10 | |
| 2310010N19Rik; regulator of G protein signaling 10; regulator of G-protein signaling 10; regulator of G-protein signalling 10; Rgs10; similar to Regulator of G-protein signaling 10 (RGS10) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54290, 6001, 67865 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43665, P49806, Q9CQE5 | |
| RGS10 | |
| A synthesized peptide derived from human RGS10(Accession O43665), corresponding to amino acid residues Q79-L129. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction