missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RGS1 293T Cell Transient Overexpression Lysate

Product Code. 18600571 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18600571 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18600571 Supplier Novus Biologicals™ Supplier No. H00005996T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 0.1 mL
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Research Category Signal Transduction
Target RGS1
Synonym 1R20, 1R20Early response protein 1R20, B-cell activation protein BL34, BL34, Early response protein 1R20, HEL-S-87, IER1, IR20, IR20immediate-early response 1, B-cell specific, regulator of G-protein signaling 1, regulator of G-protein signalling 1, RGS1
Gene Symbol RGS1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.