Learn More
Invitrogen™ RGMB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521818
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (88%), Rhesus Monkey (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Repulsive Guidance Molecule B (RGMB) is primarily expressed in the central nervous system. RGM A and B both appear at every level of the developing neural axis, where they colocalize to a large extent in the mantle layer, although only RGMA appears in the neuroepithelium, and only RGMB in the peripheral nervous system. During development, both RGMA and B appear also in lung and in limb cartilage, while RGMB has additional expression domains in pancreas.
Specifications
| RGMB | |
| Polyclonal | |
| Unconjugated | |
| RGMB | |
| 1110059F19Rik; DRAGON; DRG11-responsive axonal guidance and outgrowth of neurite; repulsive guidance molecule B; repulsive guidance molecule family member B; RGM domain family member B; RGM domain family, member B; RGMB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285704, 68799 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q6NW40, Q7TQ33 | |
| RGMB | |
| Recombinant protein encompassing a sequence within the center region of human RGMB. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.