missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: PHFPCLQKLL DYLTRMMPGS DPERRAQNLL EQFQKQEVET DNGLPNTISF SLEEEEELEG GESAEFTCFS EDLVAEQLTY MDAQLFKKVV PHHCLGCIWS RRD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
Ral GDS (Ral guanine nucleotide dissociation stimulator) is a guanine nucleotide exchange factor (GEF) that activates Ral and is implicated in oncogenic Ras-induced cell transformation. RGL1 (ral guanine nucleotide dissociation stimulator-like 1), also known as RGL or RalGDS-like 1, is a 768 amino acid protein that is a putative GEF. Strongly expressed in brain, heart, spleen, kidney and testis, RGL1 is a downstream effector protein that is involved in Ras and Ral signaling pathways. RGL1 contains an N-terminal Ras-GEF domain and a C-terminal Ras-interacting domain that interacts with the GTP-bound form of Ras through its effector loop. Due to its similarity to Ral GDS, RGL1 may be implicated in carcinogenesis. Two isoforms exist due to alternative splicing events.
Specifications
Specifications
| Antigen | RGL1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RGL1 |
| Gene Accession No. | Q9NZL6 |
| Gene Alias | KIAA0959; mKIAA0959; ral guanine nucleotide dissociation stimulator like 1; ral guanine nucleotide dissociation stimulator,-like 1; ral guanine nucleotide dissociation stimulator-like 1; RalGDS-like 1; RGL; Rgl1 |
| Gene Symbols | RGL1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
