missing translation for 'onlineSavingsMsg'
Learn More

RG3039, MedChemExpress

Product Code. 16430077
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16430077

Brand: Medchem Express HY102020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RG3039 (PF-06687859) is an orally bioavailable and brain-penetrant DcpS inhibitor with an IC50 of 0.069 nM.

RG3039 (PF-06687859) is an orally bioavailable and brain-penetrant DcpS inhibitor with an IC50 of 0.069 nM.

Specifications

Chemical Name or Material RG3039
CAS 1005504-62-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H23Cl2N5O
Quantity 50 mg
Synonym PF-06687859
Solubility Information DMSO : 5 mg/mL (11.56 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES NC1=NC(N)=C2C(OCC3CCN(CC4=C(Cl)C=CC=C4Cl)CC3)=CC=CC2=N1
Molecular Weight (g/mol) 432.35
Formula Weight 432.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.