Learn More
RG3039, MedChemExpress
Product Code. 16416427
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16416427
Brand: Medchem Express HY102020
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress RG3039 (PF-06687859) is an orally bioavailable and brain-penetrant DcpS inhibitor with an IC50 of 0.069 nM.
RG3039 (PF-06687859) is an orally bioavailable and brain-penetrant DcpS inhibitor with an IC50 of 0.069 nM.
Specifications
Chemical Name or Material RG3039
CAS 1005504-62-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H23Cl2N5O
Quantity 5 mg
Synonym PF-06687859
Solubility Information DMSO : 5 mg/mL (11.56 mM; ultrasonic and warming and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES NC1=NC(N)=C2C(OCC3CCN(CC4=C(Cl)C=CC=C4Cl)CC3)=CC=CC2=N1
Molecular Weight (g/mol) 432.35
Formula Weight 432.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation