missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ RG 108
GREENER_CHOICE

Product Code. 18054002
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18054002

Brand: Tocris Bioscience™ 3295/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-nucleoside DNA methyltransferase inhibitor

Non-nucleoside DNA methyltransferase inhibitor that blocks the enzyme active site. Inhibits DNA methylation in human cancer cell lines in vitro without detectable toxicity. Demethylates and reactivates epigenetically silenced tumor suppressor genes. Enhances the efficiency of induced pluripotent stem cell (iPS) generation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 48208-26-0
Molecular Formula C19H14N2O4

Specifications

Quantity 50 mg
Formula Weight 334.33
Purity >99%
Molecular Weight (g/mol) 334.331
Chemical Name or Material RG 108
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.