missing translation for 'onlineSavingsMsg'
Learn More

RFX3 Monoclonal Antibody (OTI3F7), TrueMAB™, OriGene

Product Code. 17545032
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17545032 30 μL 30µL
17525032 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17545032 Supplier Origene Technologies Supplier No. TA505916S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RFX3 Monoclonal antibody specifically detects RFX3 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen RFX3
Applications Western Blot
Classification Monoclonal
Clone OTI3F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Rnf39
Gene Accession No. Q9H2S5
Gene Alias bA32F11.1; C230093O12Rik; DNA binding protein RFX3; MRFX3; Regulatory factor X 3; regulatory factor X, 3 (influences HLA class II expression); regulatory factor X3; Rfx2; RFX3; Transcription factor RFX3
Host Species Mouse
Immunogen Full length human recombinant protein of human RNF39 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 386454, 80352
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.