missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RFP Polyclonal Antibody
GREENER_CHOICE

Product Code. 11528721
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11528721 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11528721 Supplier Invitrogen™ Supplier No. PA1986

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-986 detects RFP tagged proteins. This antibody is specific for the RFP tag and does not detect GFP or GFP tagged proteins. PA1-986 has been successfully used in Western blot procedures. By Western blot, this antibody detects the presence of RFP tagged proteins. PA1-986 non-specifically detects a 45 kDa protein from E. coli extracts. This non-specific cross-reactivity is not present in mammalian samples. PA1-986 immunogen is a synthetic peptide corresponding to residues V(22) N G H E F E I E G E G E G R(36) of the RFP tag. This peptide (Cat. # PEP-260) is available for use in neutralization and control experiments.

Red Fluorescent Protein (RFP) is isolated from sea anemone (Discosoma sp, Uniprot ID: Q9U6Y8), and commonly used as a molecular tag to study protein localization, reporter gene expression, and protein-protein interactions. Although it exists as a homotetramer, monomeric variants have been constructed for use in various applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Alias DsRed2; mRFP; red fluorescent; Red fluorescent protein; RFP; RFP tag
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues V(22) N G H E F E I E G E G E G R(36) of the RFP tag.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.