missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rev-erb A alpha/NR1D1 Antibody (JU34-31), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-75645
This item is not returnable.
View return policy
Description
Rev-erb A alpha/NR1D1 Monoclonal specifically detects Rev-erb A alpha/NR1D1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Rev-erb A alpha/NR1D1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
| EAR-1, EAR1nuclear receptor subfamily 1 group D member 1, HREV, nuclear receptor subfamily 1, group D, member 1, Rev-erbA-alpha, Rev-ErbAalpha, THRA1, THRALrev-erbA-alpha, V-erbA-related protein 1 | |
| Rabbit | |
| 67 kDa | |
| 100 μL | |
| 9572.0 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| JU34-31 | |
| Unconjugated | |
| TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| NR1D1 | |
| Recombinant protein within Human Rev-erb A alpha/NR1D1 aa 510-614/ 614. (SwissProt: P20393 Human; SwissProt: Q3UV55 Mouse) | |
| Protein A purified | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction