missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Retainer & Base Set (Fast) for 3500/3500xL Genetic Analyzers, 8 tube

Product Code. 15889846 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Sets
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15889846 4 Sets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15889846 Supplier Applied Biosystems™ Supplier No. 4410233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This 8-tube retainer and base set is for use with Fast 8-tube strips (0.1mL) on the 3500 series Genetic Analyzers.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This product includes 4 sets of retainers and bases.
Description Retainer & Base Set (Fast), 4 Sets, 8-well Strips, 0.1mL, Strip Tube Type, For 3500/3500xL Genetic Analyzers, Room Temperature
Format 8-well Strip
For Use With (Equipment) 3500 Genetic Analyzer, 3500xL Genetic Analyzer
For Use With (Application) Sequencing
Product Type Base Set
Quantity 4 Sets
Sample Volume 0.1 mL
Shipping Condition Room Temperature
Tube Type Strip Tube

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.