Learn More
Biochrom Resolution LITE PC Software
Product Code. 17406702
Click to view available options
Type:
Advanced PC Software
For Use With (Equipment):
Biochrom, Libra, WPA Spectrophotometers
Unit Size:
Each
Product Code. 17406702
Brand: Biochrom 80710030
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
Resolution LITE PC Software is an easy to use software platform compatible with Biochrom spectrophotometers featuring powerful modules for measurement and analysis.
Features and Benefits:
Compatible with most spectrophotometers made by Biochrom, including under the WPA and Libra sub-brands, and instruments that may have previously been supplied with Datrys software.
Key features include:
  • Quick Read and Quick Scan for fast measurements with limited setup
  • Bespoke method development for your unique laboratory requirements
  • Powerful graphical data display and manipulation options
  • Microsoft look and feel for intuitive use
  • Resolution Life Science version has pre-programmed methods in for fast measurements of commonly methods like protein and DNA/RNA/ quantification and purity measurements as well as cell density measurements and CyDye quantification
  • Library of reference material if your laboratory environment requires regular verification
  • Compatible with Windows™
  • Save, print or export your analysis as an Excel™, Extended Metafile, HTML, pdf, rich text format, text, Word or XPS file types
  • CFR version includes the ability to set up different users as part of defined groups
  • Audit trail
  • Electronic signature
Specifications
Connections PC Required
Description Spectrophotometer Analysis Software
Type Advanced PC Software
For Use With (Equipment) Biochrom, Libra, WPA Spectrophotometers