Learn More
Biochrom Resolution LITE PC Software
Click to view available options
Type:
Advanced PC Software
For Use With (Equipment):
Biochrom, Libra, WPA Spectrophotometers
Unit Size:
Each
Description
Features and Benefits:
Compatible with most spectrophotometers made by Biochrom, including under the WPA and Libra sub-brands, and instruments that may have previously been supplied with Datrys software.
Key features include:
- Quick Read and Quick Scan for fast measurements with limited setup
- Bespoke method development for your unique laboratory requirements
- Powerful graphical data display and manipulation options
- Microsoft look and feel for intuitive use
- Resolution Life Science version has pre-programmed methods in for fast measurements of commonly methods like protein and DNA/RNA/ quantification and purity measurements as well as cell density measurements and CyDye quantification
- Library of reference material if your laboratory environment requires regular verification
- Compatible with Windows™
- Save, print or export your analysis as an Excel™, Extended Metafile, HTML, pdf, rich text format, text, Word or XPS file types
- CFR version includes the ability to set up different users as part of defined groups
- Audit trail
- Electronic signature
Specifications
Specifications
| Connections | PC Required |
| Description | Spectrophotometer Analysis Software |
| Type | Advanced PC Software |
| For Use With (Equipment) | Biochrom, Libra, WPA Spectrophotometers |