Learn More
SI Analytics™ Resistance Thermometer
Product Code. 10129660 Shop All SI Analytics Products
Click to view available options
Length (Metric):
120 mm
170 mm
Model:
W 2180-KOAX
W 5790 NN
W 5791 NN
W 5980 NN
Unit Size:
Each
Product Code. 10129660
Brand: SI Analytics™ 285105254
Only null left
Add to basket
Add to basket
SI Analytics™ Resistance Thermometer is used to measure temperature and changes resistance when there is a change in temperacture.
  • Use to measure temperature
  • Comes equipt with plugs
  • Sensors Pt 1000
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Probe 4 mm
Includes 2 x 4mm banana plugs
Length (Metric) 120 mm
Model W 5790 NN
Product Type Resistance Thermometers
Description Stainless steel shaft with sensors Pt 1000
Max. Temperature (Metric) 135°C
Min. Temperature (Metric) -30°C
Temperature Range (Metric) -30°C to +135°C