Learn More
Smeg™ Resin Filler
Click to view available options
:
Capacity: 18L
Description
- Anion/cation mixed bed resins kit for the WP3000 water purifier
- Total exchange capacity 42,700 liters/°F* end of cycle at 5μS/cm (*Hardness in French degrees)
- Supplied with exhausted resins disposal bag
Specifications
Specifications
| Holds | 18L |
| Quantity | 1 Each |