Learn More
Unity Lab Services Requalification with Temperature Mapping for Shakers
Product Code. 31188983
Change view
Click to view available options
Product Type:
Service
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type
31188983 Service
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31188983 Supplier Unity Lab Services Supplier No. RQTMSHK
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Comprehensive requalification and temperature mapping for shakers and controlled chambers, verifying equipment functions and mapping temperature distribution to locate hot and cold spots.
  • Documented requalification verifying shaker performance to manufacturer specifications.
  • Key tests include door ajar, power failure recovery, and temperature alert checks.
  • Temperature mapping to identify hot and cold spots within the chamber.
  • Configurable data collection intervals with air or simulated product loads.
  • Includes open door recovery, power failure test, and detailed temperature reports.
  • Supports compliance by ensuring correct storage conditions across temperature ranges.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Service