missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies ReproTeSR™ Medium for Reprogramming (2-Component)
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- ReproTeSR™ (2-Component) is a complete and xeno-free reprogramming medium developed for generating human induced pluripotent stem (iPS) cells from fibroblasts, urine-derived cells, and blood-derived cells such as CD34+ or erythroid precursor cells under feeder-free conditions
- ReproTeSR™ is also available as part of an integrated set of tools that are optimized for reprogramming blood-derived cell types expanded in vitro from peripheral blood, such as CD34+ cells (CD34+ Progenitor Reprogramming Kit, Catalog #05925) or erythroid cells (Erythroid Progenitor Reprogramming Kit, Catalog #05924)
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells, Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Reprogramming |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction