Learn More
Brady™ Repositionable Vinyl Printer Labels
Artikelnummer. 12850099
Ansicht ändern
Click to view available options
Color:
White
Yellow
Length (Metric):
12.70
12.70 mm
15.24 m
25.40 mm
25.4mm
83.8 m
9.1 m
9.10 m
Width (Metric):
6.1 mm
8.08 mm
9.53 mm
12.70 mm
25.4 mm
38.10 mm
48.26 mm
50.8 mm
Packungsgröße:
1 roll
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Color Length (Metric) Width (Metric) unitSize
12850099 White 12.70 mm 25.4 mm Each
11509463 White 83.8 m 9.53 mm Each
11573984 Yellow 15.24 m 50.8 mm Each
11504064 Yellow 15.24 m 12.70 mm Each
11569463 White 9.1 m 12.70 mm Each
11579463 White 9.1 m 25.4 mm Each
11785663 White 25.40 mm 38.10 mm Each
11563984 White 15.24 m 50.8 mm 1 roll
11594054 White 15.24 m 12.70 mm Each
11765673 White 25.4mm 9.53 mm Each
11725683 White 12.70 38.10 mm Each
11578604 White 9.10 m 48.26 mm Each
11549463 White 83.8 m 8.08 mm Each
11508614 White 15.24 m 6.1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Artikelnummer. 12850099 Lieferant Brady™ Lieferanten-Nr. 114750
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
Repositionable vinyl cloth labels for use with Label Printer.
  • Repositionable rubber-based adhesive
  • Semi-gloss finish
Spezifikation
Certifications/Compliance RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized
Description BMP71 Label Printer Labels
Adhesive Type Rubber, Repositionable
Coating Vinyl
Color White
Finish Semi-gloss
Material Vinyl
Print Technology Thermal Transfer
Product Type Printer Label
For Use With (Application) Laboratory Identification, circuit board and component identification, wire and cable marking
For Use With (Equipment) BMP™71
Labels per Row 1
Quantity 250/Pk.
Shape Rectangle
Temperature Range (Metric) -40°C to +80°C
Thickness (Metric) 0.046 mm
Vertical Repeat (Metric) 15.88 mm
Wire Gauge 3 AWG
Dimensions (L x W) 12.70 x 25.40 mm
Length (Metric) 12.70 mm
Width (Metric) 25.4 mm
Height (Metric) 19 mm