Learn More
Thermo Scientific™ Consumable Parts for Integrated Hydride Kit for iCAP™ ICP-OES Systems
Product Code. p-7220052 Shop All Thermo Scientific Products
Click to view available options
Type:
Hydride Kit Consumables
Unit Size:
Each
This item is not returnable. View return policy
Product Code. 17691795
Brand: Thermo Scientific™ 842318050151
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Ensure peak performance of your ICP-OES analyzer with dedicated consumables for analysis of hydride-generating elements.
Ensure peak performance of your Thermo Scientific™ iCAP™ Series ICP-OES system using dedicated Thermo Scientific™ Consumable Parts for the Integrated Hydride Kit for ICP-OES systems.
Peak performance with dedicated consumables
The replacement pump tubing and a consumables kit ensures the peak performance of hydride analysis on your iCAP Series ICP-OES System.
Consumables kit contains:
  • Pump tubing (for both, mini (95 mm) and standard peristaltic pump (152 mm))
  • Teflon membrane
  • O-ring
  • Replacement tubing
  • Sample probe
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Integrated Hydride Kit for ICP-OES
Type Hydride Kit Consumables