Learn More
Thermo Scientific™ Consumable Parts for Integrated Hydride Kit for iCAP™ ICP-OES Systems
Product Code. 17691795 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Hydride Kit Consumables
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
17691795 Hydride Kit Consumables Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17691795 Supplier Thermo Scientific™ Supplier No. 842318050151
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
Ensure peak performance of your ICP-OES analyzer with dedicated consumables for analysis of hydride-generating elements.
Ensure peak performance of your Thermo Scientific™ iCAP™ Series ICP-OES system using dedicated Thermo Scientific™ Consumable Parts for the Integrated Hydride Kit for ICP-OES systems.
Peak performance with dedicated consumables
The replacement pump tubing and a consumables kit ensures the peak performance of hydride analysis on your iCAP Series ICP-OES System.
Consumables kit contains:
  • Pump tubing (for both, mini (95 mm) and standard peristaltic pump (152 mm))
  • Teflon membrane
  • O-ring
  • Replacement tubing
  • Sample probe
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Integrated Hydride Kit for ICP-OES
Type Hydride Kit Consumables