Learn More
Trajan Scientific and Medical™ Replacement SilTite™ Mini Union Ferrules
Product Code. 12957111
Click to view available options
Diameter (Metric) Inside:
0.32 mm
0.45 to 0.53 mm
Unit Size:
Pack of 10
Product Code. 12957111
Brand: Trajan Scientific and Medical™ 073472
Only null left
Add to basket
Add to basket
SilTite Mini Union delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
Specifications
Type Mini Union Ferrules
Material SilTite
Quantity 10/Pk.
Diameter (Metric) Inside 0.45 to 0.53 mm