Learn More
Trajan Scientific and Medical™ Replacement SilTite™ Mini Union Ferrules
Click to view available options
Diameter (Metric) Inside:
0.32 mm
0.45 to 0.53 mm
Unit Size:
Pack of 10
Description
- Specifically designed for connecting capillary columns
- Connects guard columns to capillary columns
- Inert and precise glass lined bore
- Low dead volume
Specifications
Specifications
| Type | Mini Union Ferrules |
| Quantity | 10/Pk. |
| Diameter (Metric) Inside | 0.32 mm |