Learn More
Hanna Instruments™ Replacement pH Electrodes
Product Code. 10619562
Change view
Click to view available options
pH Range:
0 to 14
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10619562 0 to 14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10619562 Supplier Hanna Instruments™ Supplier No. HI73127
  • ONLINE EXCLUSIVE PRICE
    167.74 €
    80.54 € / Pack of 1
    Save 87.20 €
    51% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The spare electrode is designed as an easy-fit cartridge that locks into place, featuring two o-rings to maintain a watertight seal.
  • Compatible with HI98129, and HI98130 Combo multiparameter testers, HI98127 and HI98128 pHep™ testers, and HI98121 ORP tester
  • Features a stainless steel round connector and extendable cloth junction
  • Designed as an easy-fit cartridge that locks into place, featuring two o-rings to maintain a watertight seal
Specifications
Connector Types Pin
Diameter (Metric) 12 mm
pH Range 0 to 14
Temperature (Metric) Range 0°C to 50°C